Assay Details
| Gene Symbol: Twsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023053.3 | NP_075540.1 | 4 - 5 | 645 | 92 | |
| GenBank mRNA | AF292033.1 | - | 584 | 92 | ||
| AF295097.1 | - | 3 - 4 | 503 | 92 | ||
| AJ297390.1 | - | 4 - 5 | 595 | 92 | ||
| AK049646.1 | - | 4 - 5 | 639 | 92 | ||
| AK168547.1 | - | 4 - 5 | 595 | 92 | ||
| AK168633.1 | - | 4 - 5 | 604 | 92 | ||
| BC004850.1 | - | 4 - 5 | 600 | 2810960 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Twsg1
Genomic Map