Assay Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024945.1 | NP_001020116.1 | 6 - 7 | 861 | 153 | |
| NM_023268.2 | NP_075757.1 | 6 - 7 | 861 | 153 | ||
| GenBank mRNA | AB044284.1 | - | 6 - 7 | 769 | 153 | |
| AK004880.1 | - | 6 - 7 | 807 | 153 | ||
| AK083938.1 | - | 6 - 7 | 812 | 153 | ||
| AK166839.1 | - | 6 - 7 | 807 | 153 | ||
| AK169877.1 | - | 6 - 7 | 814 | 153 | ||
| AK169920.1 | - | 6 - 7 | 810 | 153 | ||
| AK170449.1 | - | 6 - 7 | 774 | 153 | ||
| BC034131.1 | - | 3 - 4 | 353 | 153 | ||
| BC076590.1 | - | 6 - 7 | 815 | 30548238 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox1
Genomic Map