Assay Details
| Gene Symbol: Ms4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023529.2 | NP_076018.2 | 1 - 2 | 53 | 74 | |
| GenBank mRNA | AB026046.1 | - | 1 - 2 | 74 | ||
| AF237918.1 | - | 1 - 2 | 53 | 74 | ||
| BC144935.1 | - | 1 - 2 | 50 | 9053483 | 74 | |
| BC150675.1 | - | 1 - 2 | 50 | 9087586 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB026046.1,AF237918.1,BC144935.1,BC150675.1,NM_023529.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094444 | BC150675.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094444 | NM_023529.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094444 | AF237918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094444 | BC144935.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094445 | NM_023529.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094445 | AB026046.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094445 | AF237918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094445 | AK008019.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 202575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 813216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571240 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 692234 | AK039707.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294361 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63685 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177948 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123831 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094444 | BC150675.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094444 | NM_023529.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094444 | AF237918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094444 | BC144935.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094445 | NM_023529.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094445 | AB026046.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094445 | AF237918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094445 | AK008019.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P362292 | AB026046.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31953975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31954004 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123831 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177948 | - | - |
| Whole Mouse Genome G4122A | A_51_P362292 | AB026046.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138656 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123831 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177948 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14440327 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14271808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14313452 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14271812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14440328 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14271815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31954004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31954010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31954006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31954002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31953910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14271809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14429705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14433535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14385184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14333512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953920 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14333523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953912 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14491559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953915 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953916 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14433538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14433551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14194922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31954011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31953913 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10466116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10466118 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432453_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10466110 | - | - |
| Mouse Genome 430 2.0 Array | 1432453_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432453_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69826_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69826_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900010535.mm.1 | NM_023529.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010535.mm.1 | NM_023529.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010535.mm.1 | TC1900001119.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010534.mm.1 | NM_023529.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010534.mm.1 | NM_023529.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010534.mm.1 | TC1900001119.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432453_a_at | - | - |
| Mouse Expression Set 430 | 1432453_PM_a_at | - | - |
| Murine Genome U74v2 Set | 165931_PM_i_at | - | - |
| Murine Genome U74v2 Set | 165930_PM_f_at | - | - |
| Murine Genome U74v2 Set | 165930_f_at | - | - |
| Murine Genome U74v2 Set | 165931_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432453_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1432453_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432453_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432453_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 324302 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2650594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002650594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2617256 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2617256 | - | - |
| mouse-6 v1.1 (Illumina) | scl52737.9.1_36-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2650594 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52737.9.1_36-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2650594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2617256 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2617256 | - | - |

Ms4a10
Genomic Map