Assay Details
| Gene Symbol: Ms4a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023529.2 | NP_076018.2 | 4 - 5 | 426 | 75 | |
| GenBank mRNA | AB026046.1 | - | 4 - 5 | 435 | 75 | |
| AF237918.1 | - | 4 - 5 | 426 | 75 | ||
| AK008019.1 | - | 2 - 3 | 285 | 75 | ||
| BC144935.1 | - | 4 - 5 | 423 | 9053483 | 75 | |
| BC150675.1 | - | 4 - 5 | 423 | 9087586 | 75 | |
More Information
Set Membership: |
|

Ms4a10
Genomic Map