Assay Details
| Gene Symbol: Mrpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025302.3 | NP_079578.1 | 2 - 3 | 322 | 104 | |
| GenBank mRNA | AK007672.1 | - | 2 - 3 | 321 | 104 | |
| AK009521.1 | - | 2 - 3 | 316 | 104 | ||
| BC027762.1 | - | 2 - 3 | 323 | 5353459 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl2
Genomic Map