Assay Details
| Gene Symbol: Mrpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025302.3 | NP_079578.1 | 6 - 7 | 764 | 57 | |
| GenBank mRNA | AK007672.1 | - | 6 - 7 | 752 | 57 | |
| AK009521.1 | - | 6 - 7 | 758 | 57 | ||
| BC027762.1 | - | 6 - 7 | 765 | 5353459 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208080 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208078 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208079 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021886 | NM_025302.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021886 | AK007672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021886 | BC027762.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 307437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325626 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128520 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158034 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021886 | NM_025302.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021886 | AK007672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021886 | BC027762.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517138 | NM_025302.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517138 | AK007672.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517138 | AK009521.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P517138 | BC027762.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208088 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208091 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208087 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208086 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208106 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208095 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208090 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208093 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31208101 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158034 | - | - |
| Whole Mouse Genome G4122A | A_51_P517138 | NM_025302.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517138 | AK007672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517138 | AK009521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P517138 | BC027762.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118172 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158034 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224033 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13425849 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224030 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13224032 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13559159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13425880 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208079 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208078 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31208080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31208018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13256724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425872 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208103 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13224030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208099 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208095 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208090 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13425847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208093 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31208079 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445572 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445571 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445567 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445570 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445569 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445568 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445566 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448811_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445565 | - | - |
| Mouse Genome 430 2.0 Array | 1448811_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448811_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27398_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27398_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700008558.mm.1 | NM_025302.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008558.mm.1 | TC1700000898.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700008557.mm.1 | NM_025302.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008557.mm.1 | TC1700000898.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448811_PM_at | - | - |
| Mouse Expression Set 430 | 1448811_at | - | - |
| Murine Genome U74v2 Set | 161910_at | - | - |
| Murine Genome U74v2 Set | 95067_PM_at | - | - |
| Murine Genome U74v2 Set | 95067_at | - | - |
| Murine Genome U74v2 Set | 161910_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448811_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448811_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448811_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448811_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 925720 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5050161 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005050161 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5050161 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50673.7.1_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5050161 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2842366 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2842366 | - | - |

Mrpl2
Genomic Map