Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400011021.mm.1 |
NM_025544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011021.mm.1 |
NM_025544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011021.mm.1 |
TC0400001450.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011020.mm.1 |
NM_025544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011020.mm.1 |
NM_025544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011020.mm.1 |
TC0400001450.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011018.mm.1 |
NM_025544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011018.mm.1 |
NM_025544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011018.mm.1 |
TC0400001450.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011023.mm.1 |
NM_025544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011023.mm.1 |
NM_025544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011023.mm.1 |
TC0400001450.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011019.mm.1 |
NM_025544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011019.mm.1 |
NM_025544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011019.mm.1 |
TC0400001450.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400011024.mm.1 |
NM_025544.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011024.mm.1 |
NM_025544.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400011024.mm.1 |
TC0400001450.mm.1 |
MTA TC |