Assay Details
| Gene Symbol: Mrpl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025553.4 | NP_079829.1 | 1 - 2 | 208 | 72 | |
| GenBank mRNA | AB049639.1 | - | 1 - 2 | 186 | 72 | |
| AK002797.1 | - | 1 - 2 | 163 | 72 | ||
| AK010329.1 | - | 1 - 2 | 200 | 72 | ||
| AK167247.1 | - | 1 - 2 | 176 | 72 | ||
| BC046750.1 | - | 1 - 2 | 161 | 5692108 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl11
Genomic Map