Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025590.4 | NP_079866.2 | 5 - 6 | 589 | 67 | |
| GenBank mRNA | AF416923.1 | - | 5 - 6 | 527 | 67 | |
| AK031588.1 | - | 5 - 6 | 506 | 67 | ||
| AK038546.1 | - | 5 - 6 | 527 | 67 | ||
| AK048670.1 | - | 5 - 6 | 589 | 67 | ||
| AK052332.1 | - | 5 - 6 | 605 | 67 | ||
| BC042492.1 | - | 5 - 6 | 524 | 4240966 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot11
Genomic Map