Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025590.4 | NP_079866.2 | 6 - 7 | 735 | 63 | |
| GenBank mRNA | AF416923.1 | - | 6 - 7 | 673 | 63 | |
| AK031588.1 | - | 6 - 7 | 652 | 63 | ||
| AK038546.1 | - | 6 - 7 | 673 | 63 | ||
| AK048670.1 | - | 6 - 7 | 735 | 63 | ||
| AK052332.1 | - | 6 - 7 | 751 | 63 | ||
| AK122350.1 | - | 5 - 6 | 522 | 63 | ||
| BC042492.1 | - | 6 - 7 | 670 | 4240966 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot11
Genomic Map