Assay Details
| Gene Symbol: Acot11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025590.4 | NP_079866.2 | 9 - 10 | 1146 | 63 | |
| GenBank mRNA | AF416923.1 | - | 9 - 10 | 1084 | 63 | |
| AK031588.1 | - | 9 - 10 | 1063 | 63 | ||
| AK038546.1 | - | 9 - 10 | 1084 | 63 | ||
| AK048670.1 | - | 9 - 10 | 1146 | 63 | ||
| AK052332.1 | - | 9 - 10 | 1162 | 63 | ||
| AK122350.1 | - | 8 - 9 | 933 | 63 | ||
| AK170189.1 | - | 2 - 3 | 169 | 63 | ||
| AK171255.1 | - | 2 - 3 | 170 | 63 | ||
| BC042492.1 | - | 9 - 10 | 1081 | 4240966 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot11
Genomic Map