Assay Details
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026141.3 | NP_080417.2 | 3 - 4 | 261 | 99 | |
GenBank mRNA | AK011443.1 | - | 3 - 4 | 233 | 99 | |
AK012371.1 | - | 3 - 4 | 238 | 99 | ||
AK013377.1 | - | 3 - 4 | 234 | 99 | ||
AK014406.2 | - | 234 | 99 | |||
AK014453.1 | - | 3 - 4 | 249 | 99 | ||
AK017370.1 | - | 3 - 4 | 261 | 99 | ||
AK167168.1 | - | 3 - 4 | 226 | 99 | ||
AK167258.1 | - | 3 - 4 | 233 | 99 | ||
BC079912.1 | - | 3 - 4 | 223 | 30661438 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
