Assay Details
Gene Symbol: Nifk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026472.4 | NP_080748.3 | 6 - 7 | 812 | 95 | |
GenBank mRNA | AK009440.1 | - | 6 - 7 | 779 | 95 | |
AK077770.1 | - | 6 - 7 | 807 | 95 | ||
AK081486.1 | - | 6 - 7 | 779 | 95 | ||
AY030275.1 | - | 6 - 7 | 783 | 95 | ||
BC058559.1 | - | 6 - 7 | 791 | 6314031 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551221 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551219 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551220 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551218 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035205 | AK081486.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035205 | AY030275.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035205 | BB727432.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035205 | BC011356.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2035205 | BC058559.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230783 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200642 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361646 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549441 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969638 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 286114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234274 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155447 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116585 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102872 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164632 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035205 | AK081486.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035205 | AY030275.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035205 | BB727432.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035205 | BC011356.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2035205 | BC058559.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P250647 | AK081486.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P250647 | AY030275.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P250647 | BB727432.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P250647 | BC011356.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P250647 | BC058559.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551226 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551222 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551217 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551223 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20551220 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102872 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164632 | - | - |
Whole Mouse Genome G4122A | A_51_P250647 | AK081486.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P250647 | AY030275.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P250647 | BB727432.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P250647 | BC011356.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P250647 | BC058559.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113417 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102872 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164632 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00984857 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00984856 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00984854 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551221 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551220 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551218 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20551219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551304 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551293 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551312 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551307 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20551203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551244 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00996868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00763149 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551262 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551246 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00399926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00081778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00076625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551312 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551220 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551223 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00908761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551232 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00399918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20551235 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10349242 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349248 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349241 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349240 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349245 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349247 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349243 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349246 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10349244 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424001_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10349239 | - | - |
Mouse Genome 430 2.0 Array | 1424001_at | - | - |
Mouse Genome 430 2.0 Array | 1424001_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67949_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67949_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100008719.mm.1 | NM_026472.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008719.mm.1 | NM_026472.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008719.mm.1 | TC0100001058.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008717.mm.1 | NM_026472.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008717.mm.1 | NM_026472.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008717.mm.1 | TC0100001058.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100008718.mm.1 | NM_026472.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008718.mm.1 | NM_026472.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100008718.mm.1 | TC0100001058.mm.1 | MTA TC |
Affymetrix HT Mouse Genome 430A Array | 1424001_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424001_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103290563 | - | - |
MouseWG-6 v2 Expression BeadChip | 3060132 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3060132 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103290563 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103130739 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003060132 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2714936 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2714936 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1251832 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214344 | - | - |
mouse-6 v1.1 (Illumina) | scl17564.8_491-S | - | - |
mouse-6 v1.1 (Illumina) | scl0067949.2_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3060132 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103130739 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103290563 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0067949.2_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060132 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103290563 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103130739 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2714936 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2714936 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1251832 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0067949.2_13-S | - | - |
