Assay Details
| Gene Symbol: Mrps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026498.2 | NP_080774.2 | 5 - 6 | 697 | 129 | |
| GenBank mRNA | AB049945.1 | - | 5 - 6 | 421 | 129 | |
| AK002993.1 | - | 5 - 6 | 534 | 129 | ||
| AK152027.1 | - | 5 - 6 | 488 | 129 | ||
| AK160207.1 | - | 5 - 6 | 477 | 129 | ||
| AK166241.1 | - | 5 - 6 | 485 | 129 | ||
| BC005701.1 | - | 5 - 6 | 439 | 129 | ||
| BC028279.1 | - | 5 - 6 | 489 | 5370270 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | BC005701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | BC028279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | AK152027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | AK160207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | NM_026498.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | BC005701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | BC028279.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | AK152027.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | AK160207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P582394 | NM_026498.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144625 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249981 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633350 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10462 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174544 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | BC005701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | BC028279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | AK152027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | AK160207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | NM_026498.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | BC005701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | BC028279.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | AK152027.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | AK160207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P582394 | NM_026498.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25257251 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174544 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01555754 | - | - |
| Whole Mouse Genome G4122A | A_52_P582394 | BC005701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | BC028279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | AK152027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | AK160207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | NM_026498.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | BC005701.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | BC028279.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | AK152027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | AK160207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P582394 | NM_026498.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116293 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121431 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174544 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06593370 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06229311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25257356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06643739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06462030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06284147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06072282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06229311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06617325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05922986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06495701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06121545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06495694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06593371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06462027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25257290 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10554227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554228 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10554225 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460195_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455233_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10554223 | - | - |
| Mouse Genome 430 2.0 Array | 1455233_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460195_at | - | - |
| Mouse Genome 430 2.0 Array | 1460195_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455233_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67994_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67994_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700009754.mm.1 | TC0700001110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009754.mm.1 | NM_026498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009754.mm.1 | NM_026498.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009755.mm.1 | TC0700001110.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700009755.mm.1 | NM_026498.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700009755.mm.1 | NM_026498.2 | Refseq |
| Mouse Expression Set 430 | 1455233_at | - | - |
| Mouse Expression Set 430 | 1455233_PM_at | - | - |
| Mouse Expression Set 430 | 1460195_at | - | - |
| Mouse Expression Set 430 | 1460195_PM_at | - | - |
| Murine Genome U74v2 Set | 97884_PM_at | - | - |
| Murine Genome U74v2 Set | 97884_at | - | - |
| Murine Genome U74v2 Set | 169810_PM_at | - | - |
| Murine Genome U74v2 Set | 169810_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460195_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460195_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455233_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455233_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455233_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460195_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460195_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455233_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 906948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3130270 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6660735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006660735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003130270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2681946 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2659082 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2681946 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2659082 | - | - |
| mouse-6 v1.1 (Illumina) | scl00036.1_15-S | - | - |
| mouse-6 v1.1 (Illumina) | scl067994.6_157-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6660735 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3130270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00036.1_15-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067994.6_157-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6660735 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3130270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2681946 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2786708 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2786708 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2681946 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00036.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067994.6_157-S | - | - |

Mrps11
Genomic Map