Assay Details
Gene Symbol: Sycn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026716.3 | NP_080992.3 | 1 - 2 | 421 | 61 | |
GenBank mRNA | AK003083.1 | - | 1 - 2 | 429 | 61 | |
BC019567.1 | - | 1 - 2 | 421 | 4208065 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963639 | NM_026716.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963639 | AK003083.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1963639 | BC019567.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721000 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963639 | NM_026716.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963639 | AK003083.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1963639 | BC019567.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022372 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022406 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022398 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022389 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022377 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022373 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022379 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022407 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022388 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022383 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022402 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022405 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022384 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022381 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022386 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022375 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022387 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022391 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022378 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022401 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25022410 | - | - |
Whole Mouse Genome G4122A | A_51_P434332 | NM_026716.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P434332 | AK003083.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P434332 | BC019567.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123504 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05978371 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06582348 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06154376 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06582349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022424 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022461 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022408 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022448 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022393 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022451 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022439 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022444 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022441 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022420 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022433 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25022417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06582358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022402 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05986887 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06582341 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022355 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022373 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06443835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022358 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06154376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05894959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06332985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022397 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022372 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022359 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25022388 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10551532 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551533 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434803_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451228_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10551531 | - | - |
Mouse Genome 430 2.0 Array | 1434803_a_at | - | - |
Mouse Genome 430 2.0 Array | 1451228_a_at | - | - |
Mouse Genome 430 2.0 Array | 1451228_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1434803_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68416_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68416_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700004103.mm.1 | TC0700000505.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004103.mm.1 | NM_026716.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004103.mm.1 | NM_026716.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004104.mm.1 | TC0700000505.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700004104.mm.1 | NM_026716.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700004104.mm.1 | NM_026716.3 | Refseq |
Mouse Expression Set 430 | 1434803_a_at | - | - |
Mouse Expression Set 430 | 1451228_PM_a_at | - | - |
Mouse Expression Set 430 | 1434803_PM_a_at | - | - |
Mouse Expression Set 430 | 1451228_a_at | - | - |
Murine Genome U74v2 Set | 95509_PM_at | - | - |
Murine Genome U74v2 Set | 95509_at | - | - |
Mouse Genome 430A 2.0 Array | 1451228_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1434803_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1434803_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451228_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434803_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451228_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451228_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434803_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 654297 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2570040 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2570040 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002570040 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232533 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1232533 | - | - |
mouse-6 v1.1 (Illumina) | scl068416.1_61-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2570040 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl068416.1_61-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570040 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1232533 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1232533 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl068416.1_61-S | - | - |
