Assay Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110274.1 | NP_001103744.1 | 3 - 4 | 507 | 70 | |
| NM_030683.3 | NP_109608.1 | 7 - 8 | 1133 | 70 | ||
| NM_207651.3 | NP_997534.2 | 7 - 8 | 1133 | 70 | ||
| GenBank mRNA | AF258601.2 | - | 3 - 4 | 507 | 70 | |
| AF258602.1 | - | 7 - 8 | 1034 | 70 | ||
| AF366052.2 | - | 7 - 8 | 1125 | 70 | ||
| AY221737.1 | - | 2 - 3 | 219 | 70 | ||
| BC150680.1 | - | 7 - 8 | 1041 | 9087591 | 70 | |
More Information
Set Membership: |
|

Slc14a2
Genomic Map