Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
NM_001110274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
NM_001110274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
NM_030683.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
NM_030683.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
NM_207651.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
NM_207651.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011792.mm.1 |
TC1800001638.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011790.mm.1 |
NM_001110274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011790.mm.1 |
NM_001110274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011790.mm.1 |
NM_030683.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011790.mm.1 |
NM_030683.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011790.mm.1 |
TC1800001638.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
NM_001110274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
NM_001110274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
NM_030683.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
NM_030683.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
NM_207651.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
NM_207651.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800011793.mm.1 |
TC1800001638.mm.1 |
MTA TC |