Assay Details
| Gene Symbol: Ucn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031250.5 | NP_112540.2 | 2 | 666 | 67 | |
| GenBank mRNA | AK148451.1 | - | 2 | 667 | 67 | |
| BC119433.1 | - | 1 | 151 | 40041175 | 67 | |
| BC119434.1 | - | 1 | 148 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to stress
adenylate cyclase-activating G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
digestion
cellular response to nutrient levels
positive regulation of insulin secretion
positive regulation of membrane potential
G-protein coupled receptor binding
hormone activity
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone receptor 2 binding
response to stress
adenylate cyclase-activating G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
digestion
cellular response to nutrient levels
positive regulation of insulin secretion
positive regulation of membrane potential
G-protein coupled receptor binding
hormone activity
neuropeptide hormone activity
protein binding
corticotropin-releasing hormone receptor 2 binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046403 | AK148451.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046403 | NM_031250.5 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174096 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105954 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046403 | AK148451.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046403 | NM_031250.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342327 | AK148451.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342327 | BC119433.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342327 | BC119434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342327 | NM_031250.5 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02619568 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105954 | - | - |
| Whole Mouse Genome G4122A | A_51_P342327 | AK148451.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342327 | BC119433.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342327 | BC119434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342327 | NM_031250.5 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107170 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105954 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10959610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10878788 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10959606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28790699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10898905 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10683145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11067643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10630984 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10604074 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10605001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790750 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790765 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790721 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10803237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10959612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10898907 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10881300 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10476551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10683142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10967971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10847753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790763 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28790768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10556517 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10407432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10407434 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441439_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10407431 | - | - |
| Mouse Genome 430 2.0 Array | 1441439_at | - | - |
| Mouse Genome 430 2.0 Array | 1450607_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1441439_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1450607_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83428_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83428_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300008625.mm.1 | NM_031250.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008625.mm.1 | NM_031250.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008625.mm.1 | TC1300001375.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300008623.mm.1 | NM_031250.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008623.mm.1 | NM_031250.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300008623.mm.1 | TC1300001375.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450607_PM_s_at | - | - |
| Mouse Expression Set 430 | 1450607_s_at | - | - |
| Mouse Expression Set 430 | 1441439_PM_at | - | - |
| Mouse Expression Set 430 | 1441439_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450607_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450607_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441439_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441439_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 538528 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4920020 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4920020 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920020 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1238143 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1238143 | - | - |
| mouse-6 v1.1 (Illumina) | scl44315.2.7_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920020 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44315.2.7_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920020 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1238143 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238143 | - | - |

Ucn3
Genomic Map