Assay Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032398.2 | NP_115774.2 | 3 - 4 | 1222 | 85 | |
| GenBank mRNA | AF348403.1 | - | 3 - 4 | 1193 | 85 | |
| AF369900.1 | - | 3 - 4 | 1217 | 85 | ||
| BC013517.1 | - | 3 - 4 | 1225 | 4235957 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plvap
Genomic Map