Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800002777.mm.1 |
NM_033595.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002777.mm.1 |
NM_033595.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002777.mm.1 |
TC1800001741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800002778.mm.1 |
NM_033595.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002778.mm.1 |
NM_033595.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002778.mm.1 |
TC1800001741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800002776.mm.1 |
NM_033595.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002776.mm.1 |
NM_033595.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002776.mm.1 |
TC1800001741.mm.1 |
MTA TC |