Assay Details
| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080638.3 | NP_542369.2 | 6 - 7 | 778 | 72 | |
| GenBank mRNA | AF210456.1 | - | 6 - 7 | 780 | 72 | |
| AK088046.1 | - | 6 - 7 | 763 | 72 | ||
| AK152534.1 | - | 6 - 7 | 753 | 72 | ||
| AK167899.1 | - | 6 - 7 | 754 | 72 | ||
| AK168100.1 | - | 6 - 7 | 754 | 72 | ||
| BC006709.1 | - | 6 - 7 | 743 | 3584439 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvp
Genomic Map