Assay Details
| Gene Symbol: Mvp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080638.3 | NP_542369.2 | 12 - 13 | 2239 | 69 | |
| GenBank mRNA | AF210456.1 | - | 12 - 13 | 2241 | 69 | |
| AK019080.1 | - | 2 - 3 | 434 | 69 | ||
| AK088046.1 | - | 12 - 13 | 2224 | 69 | ||
| AK167899.1 | - | 12 - 13 | 2215 | 69 | ||
| AK168100.1 | - | 12 - 13 | 2215 | 69 | ||
| BC006709.1 | - | 12 - 13 | 2204 | 3584439 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mvp
Genomic Map