Assay Details
| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163776.1 | NP_001157248.1 | 8 - 9 | 973 | 85 | |
| NM_080727.2 | NP_542765.2 | 7 - 8 | 833 | 85 | ||
| GenBank mRNA | AJ300738.1 | - | 7 - 8 | 833 | 85 | |
| AK158203.1 | - | 8 - 9 | 976 | 85 | ||
| BC112375.1 | - | 7 - 8 | 830 | 40057032 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss3
Genomic Map