Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
NM_001037759.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
NM_001037759.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
NM_133220.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
NM_133220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
NM_177547.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
NM_177547.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000363.mm.1 |
TC0100000044.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
NM_001037759.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
NM_001037759.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
NM_133220.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
NM_133220.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
NM_177547.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
NM_177547.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100000364.mm.1 |
TC0100000044.mm.1 |
MTA TC |