Assay Details
| Gene Symbol: Mlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133241.2 | NP_573504.1 | 3 - 4 | 326 | 71 | |
| GenBank mRNA | AF449425.1 | - | 3 - 4 | 780 | 71 | |
| AK030396.1 | - | 3 - 4 | 325 | 71 | ||
| AK043757.1 | - | 3 - 4 | 326 | 71 | ||
| AK122190.1 | - | 3 - 4 | 768 | 71 | ||
| BC024719.1 | - | 3 - 4 | 326 | 5363238 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlc1
Genomic Map