Assay Details
| Gene Symbol: Tubgcp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286007.1 | NP_001272936.1 | 15 - 16 | 2651 | 66 | |
| NM_001286011.1 | NP_001272940.1 | 14 - 15 | 2418 | 66 | ||
| NM_133755.2 | NP_598516.2 | 15 - 16 | 2578 | 66 | ||
| GenBank mRNA | AK006233.1 | - | 5 - 6 | 680 | 66 | |
| AK146771.1 | - | 15 - 16 | 2573 | 66 | ||
| AK158060.1 | - | 15 - 16 | 2367 | 66 | ||
| AK167924.1 | - | 15 - 16 | 2473 | 66 | ||
| BC012519.1 | - | 15 - 16 | 2506 | 3601589 | 66 | |
| BC025564.1 | - | 14 - 15 | 2340 | 66 | ||
| BC025582.1 | - | 15 - 16 | 2467 | 5324981 | 66 | |
| BC027034.1 | - | 14 - 15 | 2852 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp2
Genomic Map