Assay Details
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286007.1 | NP_001272936.1 | 16 - 17 | 2786 | 83 | |
NM_001286011.1 | NP_001272940.1 | 15 - 16 | 2553 | 83 | ||
NM_133755.2 | NP_598516.2 | 16 - 17 | 2713 | 83 | ||
GenBank mRNA | AK006233.1 | - | 6 - 7 | 815 | 83 | |
AK146771.1 | - | 16 - 17 | 2708 | 83 | ||
AK158060.1 | - | 16 - 17 | 2502 | 83 | ||
AK167924.1 | - | 16 - 17 | 2608 | 83 | ||
BC012519.1 | - | 16 - 17 | 2641 | 3601589 | 83 | |
BC025564.1 | - | 15 - 16 | 2475 | 83 | ||
BC025582.1 | - | 16 - 17 | 2602 | 5324981 | 83 | |
BC027034.1 | - | 15 - 16 | 2987 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
