Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900010472.mm.1 |
NM_133804.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010472.mm.1 |
NM_133804.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010472.mm.1 |
TC1900001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010471.mm.1 |
NM_133804.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010471.mm.1 |
NM_133804.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010471.mm.1 |
TC1900001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010469.mm.1 |
NM_133804.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010469.mm.1 |
NM_133804.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010469.mm.1 |
TC1900001115.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900010470.mm.1 |
NM_133804.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010470.mm.1 |
NM_133804.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010470.mm.1 |
TC1900001115.mm.1 |
MTA TC |