Assay Details
Gene Symbol: Ptov1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133949.1 | NP_598710.1 | 6 - 7 | 983 | 103 | |
GenBank mRNA | AK188193.1 | - | 2 - 3 | 179 | 103 | |
AK211594.1 | - | 2 - 3 | 286 | 103 | ||
BC008543.1 | - | 6 - 7 | 859 | 3496037 | 103 | |
BC024632.1 | - | 6 - 7 | 983 | 4222241 | 103 | |
BC062108.1 | - | 1 - 2 | 1305 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
