Assay Details
Gene Symbol: Vmn1r230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134197.1 | NP_598958.1 | 1 | 895 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2124258 | NM_134197.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 708334 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105077 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2124258 | NM_134197.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P182658 | NM_134197.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084028 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084030 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084053 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084050 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084023 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084037 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084052 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084046 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084040 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084009 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084006 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084001 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084012 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084008 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084018 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084011 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31083999 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31084016 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105077 | - | - |
Whole Mouse Genome G4122A | A_51_P182658 | NM_134197.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120330 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105077 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505034 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505035 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505041 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13453930 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505038 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13453916 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13505036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31083996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31083993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31083999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31083991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31083994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31083989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31084009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084018 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084034 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084028 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084011 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084006 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13281561 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13453929 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084001 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084031 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084051 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084037 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084038 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084046 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084041 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084012 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13182891 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13420858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083993 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084056 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084040 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13281558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084008 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084057 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13505035 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084014 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084025 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084054 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13096421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084021 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084013 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084047 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13182893 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084049 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084026 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084044 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083989 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084016 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084009 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31084004 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31083991 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10442152 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10442151 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700001874.mm.1 | NM_134197.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700001874.mm.1 | NM_134197.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700001874.mm.1 | TC1700000253.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 527210 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4480253 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4480253 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004480253 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217421 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217421 | - | - |
mouse-6 v1.1 (Illumina) | scl8633.1.1_106-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4480253 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl8633.1.1_106-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480253 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217421 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217421 | - | - |
