Assay Details
| Gene Symbol: Vmn1r212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134241.1 | NP_599002.1 | 1 | 941 | 101 | |
| GenBank mRNA | BC132251.1 | - | 1 | 985 | 40130528 | 101 |
| BC132253.1 | - | 1 | 985 | 40130530 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874758 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139562 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879104 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879092 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879096 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879083 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879089 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879085 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879080 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879076 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28879094 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139562 | - | - |
| Whole Mouse Genome G4122A | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433650 | BC132251.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433650 | BC132253.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P433650 | NM_134241.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109074 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28879120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10984530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879075 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10679688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10888637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879112 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879104 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879089 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879113 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879116 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879108 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879109 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879102 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879119 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10984558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879032 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10888640 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10984559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28879080 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10408155 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10408154 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300009988.mm.1 | TC1300001609.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009989.mm.1 | NM_134241.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009989.mm.1 | NM_134241.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300009989.mm.1 | TC1300001609.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300009990.mm.1 | TC1300001609.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 340863 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103610128 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103610128 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360059 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360059 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243438 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219757 | - | - |
| mouse-6 v1.1 (Illumina) | scl11513.1.1_216-S | - | - |
| mouse-6 v1.1 (Illumina) | GI_21717784-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103610128 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360059 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl11513.1.1_216-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360059 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103610128 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1219757 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219757 | - | - |

Vmn1r212
Genomic Map