Assay Details
| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134250.2 | NP_599011.2 | 3 - 4 | 548 | 85 | |
| GenBank mRNA | AF399831.1 | - | 3 - 4 | 494 | 85 | |
| AF450241.1 | - | 3 - 4 | 544 | 85 | ||
| AK148474.1 | - | 3 - 4 | 549 | 85 | ||
| AK170775.1 | - | 4 - 5 | 368 | 85 | ||
| AY553334.1 | - | 3 - 4 | 541 | 85 | ||
| BC106851.2 | - | 3 - 4 | 506 | 40046221 | 85 | |
| BC106852.2 | - | 3 - 4 | 506 | 40046222 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Havcr2
Genomic Map