Assay Details
| Gene Symbol: Trpm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134252.3 | NP_599013.1 | 14 - 15 | 1931 | 62 | |
| GenBank mRNA | AF481480.2 | - | 2200 | 62 | ||
| AY095352.1 | - | 12 - 13 | 1765 | 62 | ||
| BC117934.1 | - | 14 - 15 | 1931 | 40097778 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpm8
Genomic Map