Assay Details
| Gene Symbol: Olr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138648.2 | NP_619589.2 | 3 - 4 | 473 | 79 | |
| GenBank mRNA | AF303744.1 | - | 3 - 4 | 462 | 79 | |
| AK154687.1 | - | 3 - 4 | 474 | 79 | ||
| AK160919.1 | - | 3 - 4 | 473 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Olr1
Genomic Map