Assay Details
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139200.4 | NP_631939.1 | 3 - 4 | 343 | 89 | |
GenBank mRNA | AF192525.1 | - | 3 - 4 | 263 | 89 | |
AK089364.1 | - | 3 - 4 | 284 | 89 | ||
AK152784.1 | - | 3 - 4 | 325 | 89 | ||
AK157859.1 | - | 3 - 4 | 344 | 89 | ||
AK164790.1 | - | 4 - 5 | 349 | 89 | ||
AK166254.1 | - | 3 - 4 | 344 | 89 | ||
AK168343.1 | - | 3 - 4 | 343 | 89 | ||
AK169575.1 | - | 3 - 4 | 329 | 89 | ||
AK170544.1 | - | 3 - 4 | 344 | 89 | ||
AK170796.1 | - | 3 - 4 | 344 | 89 | ||
AK171467.1 | - | 3 - 4 | 344 | 89 | ||
AK172380.1 | - | 3 - 4 | 344 | 89 | ||
BC007144.1 | - | 3 - 4 | 332 | 3487179 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
