Assay Details
| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145138.2 | NP_660120.2 | 9 - 10 | 1127 | 60 | |
| GenBank mRNA | AJ489828.1 | - | 9 - 10 | 1102 | 60 | |
| AK173323.1 | - | 9 - 10 | 1111 | 60 | ||
| BC117971.2 | - | 9 - 10 | 1068 | 40098375 | 60 | |
| BC117972.2 | - | 9 - 10 | 1067 | 40098378 | 60 | |
More Information
Set Membership: |
|

Nek9
Genomic Map