Assay Details
| Gene Symbol: Sds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145565.1 | NP_663540.1 | 2 - 3 | 206 | 82 | |
| GenBank mRNA | AK149438.1 | - | 2 - 3 | 223 | 82 | |
| BC011259.1 | - | 2 - 3 | 203 | 82 | ||
| BC021605.1 | - | 2 - 3 | 203 | 5097741 | 82 | |
| BC021950.1 | - | 2 - 3 | 206 | 5102037 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sds
Genomic Map