Assay Details
| Gene Symbol: Sds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145565.1 | NP_663540.1 | 6 - 7 | 706 | 61 | |
| GenBank mRNA | AK149438.1 | - | 6 - 7 | 723 | 61 | |
| BC011259.1 | - | 6 - 7 | 703 | 61 | ||
| BC021605.1 | - | 6 - 7 | 703 | 5097741 | 61 | |
| BC021950.1 | - | 6 - 7 | 706 | 5102037 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
gluconeogenesis
cellular amino acid metabolic process
L-serine catabolic process
threonine catabolic process
pyruvate biosynthetic process
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
lyase activity
pyridoxal phosphate binding
protein homodimerization activity
protein dimerization activity
gluconeogenesis
cellular amino acid metabolic process
L-serine catabolic process
threonine catabolic process
pyruvate biosynthetic process
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
lyase activity
pyridoxal phosphate binding
protein homodimerization activity
protein dimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853117 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856915 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688618 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327731 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48947 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102385 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161540 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24038925 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102385 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161540 | - | - |
| Whole Mouse Genome G4122A | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | NM_145565.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | AK149438.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | BC011259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | BC021605.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P249335 | BC021950.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138424 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102385 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161540 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04761491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24039021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24038950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04761491 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04761492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038974 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04125812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04761495 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04761493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04290714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874281 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874282 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04874286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04491964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04761500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04405299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04776525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24038966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04938635 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525079 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525081 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525078 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525077 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424744_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525076 | - | - |
| Mouse Genome 430 2.0 Array | 1424744_at | - | - |
| Mouse Genome 430 2.0 Array | 1424744_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231691_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231691_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500011646.mm.1 | TC0500001337.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011644.mm.1 | TC0500001337.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011644.mm.1 | NM_145565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011644.mm.1 | NM_145565.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011645.mm.1 | TC0500001337.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500011645.mm.1 | NM_145565.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500011645.mm.1 | NM_145565.1 | Refseq |
| Mouse Expression Set 430 | 1424744_at | - | - |
| Mouse Expression Set 430 | 1424744_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424744_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424744_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424744_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424744_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 920177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 110242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 110242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6380725 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110242 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006380725 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255497 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255497 | - | - |
| mouse-6 v1.1 (Illumina) | scl0004108.1_20-S | - | - |
| mouse-6 v1.1 (Illumina) | scl27295.8.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6380725 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110242 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27295.8.1_2-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0004108.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110242 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380725 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255497 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2763983 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255497 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0004108.1_20-S | - | - |

Sds
Genomic Map