Assay Details
Gene Symbol: Nrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147201.2 | NP_671734.1 | 16 - 17 | 1548 | 50 | |
GenBank mRNA | AF302138.1 | - | 16 - 17 | 1547 | 50 | |
AK046142.1 | - | 17 - 18 | 1566 | 50 | ||
AK077440.1 | - | 16 - 17 | 1585 | 50 | ||
AK170501.1 | - | 16 - 17 | 1564 | 50 | ||
AK219460.1 | - | 5 - 6 | 328 | 50 | ||
BC004756.1 | - | 16 - 17 | 1518 | 3154089 | 50 | |
BC013212.1 | - | 5 - 6 | 942 | 50 | ||
BC018463.1 | - | 16 - 17 | 1522 | 3598438 | 50 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
