Assay Details
| Gene Symbol: Plcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081456.1 | NP_001074925.1 | 3 - 4 | 355 | 71 | |
| NM_148937.2 | NP_683739.2 | 3 - 4 | 506 | 71 | ||
| GenBank mRNA | AK016945.1 | - | 3 - 4 | 503 | 71 | |
| AK029561.1 | - | 3 - 4 | 501 | 71 | ||
| AK039149.1 | - | 3 - 4 | 333 | 71 | ||
| AK140231.1 | - | 3 - 4 | 342 | 71 | ||
| AY033991.1 | - | 3 - 4 | 338 | 71 | ||
| BC066156.1 | - | 3 - 4 | 246 | 30099811 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcd4
Genomic Map