Assay Details
Gene Symbol: Bgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007542.4 | NP_031568.2 | 1 - 2 | 129 | 72 | |
GenBank mRNA | AK086920.1 | - | 1 - 2 | 126 | 72 | |
AK135496.1 | - | 1 - 2 | 130 | 72 | ||
AK155973.1 | - | 1 - 2 | 130 | 72 | ||
AK164867.1 | - | 1 - 2 | 130 | 72 | ||
AK171870.1 | - | 1 - 2 | 130 | 72 | ||
BC005452.1 | - | 1 - 2 | 123 | 3594469 | 72 | |
BC019502.1 | - | 1 - 2 | 132 | 4237268 | 72 | |
BC052857.1 | - | 1 - 2 | 114 | 30061850 | 72 | |
BC057185.1 | - | 1 - 2 | 141 | 72 | ||
L20276.1 | - | 1 - 2 | 129 | 72 | ||
X53928.1 | - | 224 | 72 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK135496.1,AK155973.1,AK164867.1,AK171870.1,BC005452.1,BC019502.1,BC052857.1,BC057185.1,L20276.1,NM_007542.4,X53928.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
