Assay Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177647.2 | NP_001171118.1 | 1 - 2 | 528 | 83 | |
| NM_001291019.1 | NP_001277948.1 | 1 - 2 | 528 | 83 | ||
| NM_001291020.1 | NP_001277949.1 | 2 - 3 | 330 | 83 | ||
| NM_001291021.1 | NP_001277950.1 | 1 - 2 | 528 | 83 | ||
| NM_007547.4 | NP_031573.2 | 1 - 2 | 528 | 83 | ||
| GenBank mRNA | AB018194.1 | - | 1 - 2 | 94 | 83 | |
| AF072543.1 | - | 1 - 2 | 84 | 83 | ||
| AF072544.1 | - | 1 - 2 | 84 | 83 | ||
| AF332080.1 | - | 1 - 2 | 94 | 83 | ||
| AK159617.1 | - | 2 - 3 | 331 | 83 | ||
| BC062197.1 | - | 1 - 2 | 119 | 5368250 | 83 | |
| BQ770331.1 | - | 1 - 2 | 502 | 83 | ||
| CD578948.1 | - | 1 - 2 | 528 | 83 | ||
| CX236057.1 | - | 1 - 2 | 113 | 83 | ||
| D85785.1 | - | 1 - 2 | 83 | |||
| U89694.1 | - | 1 - 2 | 105 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sirpa
Genomic Map