Assay Details
| Gene Symbol: Ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009212.2 | NP_033238.2 | 2 - 3 | 483 | 85 | |
| GenBank mRNA | AK142456.1 | - | 2 - 3 | 287 | 85 | |
| AK153763.1 | - | 2 - 3 | 444 | 85 | ||
| AK154805.1 | - | 2 - 3 | 480 | 85 | ||
| AK155899.1 | - | 2 - 3 | 480 | 85 | ||
| BC052025.1 | - | 2 - 3 | 453 | 85 | ||
| BC060085.1 | - | 2 - 3 | 453 | 85 | ||
| BC066215.1 | - | 2 - 3 | 426 | 85 | ||
| BC083142.1 | - | 2 - 3 | 288 | 85 | ||
| BC133681.1 | - | 2 - 3 | 291 | 85 | ||
| L10075.1 | - | 2 - 3 | 379 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ighmbp2
Genomic Map