Assay Details
| Gene Symbol: Zfp143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009281.3 | NP_033307.2 | 3 - 4 | 284 | 63 | |
| GenBank mRNA | AF011758.2 | - | 230 | 63 | ||
| AK028056.1 | - | 3 - 4 | 274 | 63 | ||
| AK029078.1 | - | 3 - 4 | 276 | 63 | ||
| AK029926.1 | - | 3 - 4 | 549 | 63 | ||
| BC037658.1 | - | 3 - 4 | 275 | 5356405 | 63 | |
| CN709211.1 | - | 3 - 4 | 284 | 63 | ||
More Information
Set Membership: |
|

Zfp143
Genomic Map