Assay Details
| Gene Symbol: Zfp143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009281.3 | NP_033307.2 | 5 - 6 | 443 | 62 | |
| GenBank mRNA | AF011758.2 | - | 392 | 62 | ||
| AK028056.1 | - | 5 - 6 | 436 | 62 | ||
| AK029078.1 | - | 5 - 6 | 435 | 62 | ||
| AK029926.1 | - | 5 - 6 | 708 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp143
Genomic Map