Assay Details
| Gene Symbol: Tpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009417.2 | NP_033443.1 | 6 - 7 | 662 | 79 | |
| GenBank mRNA | AK047843.1 | - | 6 - 7 | 641 | 79 | |
| X60703.1 | - | 6 - 7 | 662 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpo
Genomic Map