Assay Details
| Gene Symbol: Adam17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277266.1 | NP_001264195.1 | 9 - 10 | 1208 | 91 | |
| NM_001291871.1 | NP_001278800.1 | 8 - 9 | 1170 | 91 | ||
| NM_009615.6 | NP_033745.4 | 8 - 9 | 1151 | 91 | ||
| NR_102380.1 | 5 - 6 | 794 | 91 | |||
| NR_120376.1 | 8 - 9 | 1151 | 91 | |||
| GenBank mRNA | AB021709.1 | - | 8 - 9 | 1112 | 91 | |
| AJ007365.1 | - | 8 - 9 | 951 | 91 | ||
| AK137693.1 | - | 8 - 9 | 1058 | 91 | ||
| AK139471.1 | - | 8 - 9 | 1057 | 91 | ||
| AK142441.1 | - | 8 - 9 | 1110 | 91 | ||
| AK144161.1 | - | 9 - 10 | 1138 | 91 | ||
| AK149288.1 | - | 8 - 9 | 1054 | 91 | ||
| AK149614.1 | - | 8 - 9 | 1058 | 91 | ||
| BC026052.1 | - | 5 - 6 | 690 | 91 | ||
| BC048792.1 | - | 8 - 9 | 1058 | 91 | ||
| BC094655.1 | - | 8 - 9 | 1065 | 5705503 | 91 | |
| BC114999.1 | - | 8 - 9 | 1011 | 6506501 | 91 | |
| BC138416.1 | - | 8 - 9 | 1030 | 8861436 | 91 | |
| BC138420.1 | - | 8 - 9 | 1030 | 8861440 | 91 | |
| BC145270.1 | - | 8 - 9 | 1030 | 9053824 | 91 | |
| U69613.1 | - | 8 - 9 | 1019 | 91 | ||
| U69614.1 | - | 8 - 9 | 1103 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adam17
Genomic Map