Assay Details
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 10 - 11 | 992 | 63 | |
GenBank mRNA | BC060975.1 | - | 10 - 11 | 955 | 63 | |
BC129920.1 | - | 10 - 11 | 958 | 63 | ||
X64227.1 | - | 10 - 11 | 1002 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
