Assay Details
Gene Symbol: Parp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009632.2 | NP_033762.1 | 1 - 2 | 43 | 82 | |
GenBank mRNA | AJ007780.1 | - | 1 - 2 | 56 | 82 | |
AK076193.1 | - | 1 - 2 | 70 | 82 | ||
AK141876.1 | - | 1 - 2 | 65 | 82 | ||
BC052000.1 | - | 1 - 2 | 43 | 82 | ||
BC062150.1 | - | 1 - 2 | 43 | 6333875 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
