Assay Details
| Gene Symbol: Snap25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011428.3 | NP_035558.1 | 5 - 6 | 496 | 90 | |
| GenBank mRNA | AF483516.1 | - | 4 - 5 | 283 | 90 | |
| AF483517.1 | - | 4 - 5 | 283 | 90 | ||
| AK078038.1 | - | 5 - 6 | 494 | 90 | ||
| BC018249.1 | - | 5 - 6 | 384 | 4504644 | 90 | |
| M22012.1 | - | 5 - 6 | 446 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snap25
Genomic Map